Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
thul16
Entrez Gene ID
496862
Gene Name
thyroid hormone up-regulated protein (gene 16)
Gene Aliases
wd40
Chromosome Location
Chr.8: 67751444 - 67764601 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011392.1 NP_001011392.1 12 - 13 1650 103
XM_018096243.1 XP_017951732.1 12 - 13 1175 103
XM_018096244.1 XP_017951733.1 12 - 13 1417 103
GenBank mRNA BC088584.1 - 12 - 13 1650 103

View Full Product Details