Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
serinc1
Entrez Gene ID
496872
Gene Name
serine incorporator 1
Gene Aliases
serinc1-a, serinc1-b, tde1l, tde2, tms-2
Chromosome Location
Chr.5: 68109422 - 68121311 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011399.1 NP_001011399.1 5 - 6 740 80
XM_018093869.1 XP_017949358.1 5 - 6 700 80
GenBank mRNA BC088606.1 - 5 - 6 740 80

View Full Product Details