Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hoxa5
Entrez Gene ID
496880
Gene Name
homeobox A5
Gene Aliases
hox-a5, hox1, hox1.3, hox1c
Chromosome Location
Chr.6: 32918430 - 32920666 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011405.1 NP_001011405.1 1 - 2 633 67
GenBank mRNA BC088772.1 - 1 - 2 633 67

View Full Product Details