Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eef1a2
Entrez Gene ID
496898
Gene Name
eukaryotic translation elongation factor 1 alpha 2
Gene Aliases
eef1al, ef1a, stnl
Chromosome Location
Chr.Un|NW_016683902: 87 - 8651 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011418.1 NP_001011418.1 1 - 2 180 76
GenBank mRNA BC088010.1 - 1 - 2 180 76

View Full Product Details