Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- hsbp1
- Entrez Gene ID
-
496903
- Gene Name
- heat shock factor binding protein 1
- Gene Aliases
-
hsbp1-a, hsbp1-b, npc-a-13
- Chromosome Location
-
Chr.4: 65020463 - 65026074 on Build UCB_Xtro_10.0
- Gene Symbol
- sucla2
- Entrez Gene ID
-
394842
- Gene Name
- succinate-CoA ligase, ADP-forming, beta subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 131528806 - 131553326 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CR855637.2 |
- |
- |
1575 |
95 |
View Full Product Details