Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wdr20
Entrez Gene ID
496935
Gene Name
WD repeat domain 20
Gene Aliases
-
Chromosome Location
Chr.8: 76601237 - 76636991 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011447.1 NP_001011447.1 2 - 3 446 91
XM_012968324.2 XP_012823778.1 2 - 3 693 91
GenBank mRNA BC088535.1 - 2 - 3 446 91
CR848483.2 - 2 - 3 476 91

View Full Product Details