Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pnpla3
Entrez Gene ID
548410
Gene Name
patatin like phospholipase domain containing 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683517: 152125 - 170631 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015693.1 NP_001015693.1 8 - 9 1222 86
GenBank mRNA BC089233.1 - 8 - 9 1222 86

View Full Product Details