Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp2d6
Entrez Gene ID
548436
Gene Name
cytochrome P450 family 2 subfamily D member 6
Gene Aliases
cyp2d2
Chromosome Location
Chr.4: 109819977 - 109830963 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015719.1 NP_001015719.1 5 - 6 1060 110
GenBank mRNA BC089651.1 - 5 - 6 1060 110

View Full Product Details