Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
serpini2
Entrez Gene ID
548471
Gene Name
serpin peptidase inhibitor, clade I (pancpin), member 2
Gene Aliases
-
Chromosome Location
Chr.5: 101585732 - 101609372 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015754.1 NP_001015754.1 6 - 7 1023 116
GenBank mRNA BC089690.1 - 6 - 7 1023 116

View Full Product Details