Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rasl11b
Entrez Gene ID
548491
Gene Name
RAS like family 11 member B
Gene Aliases
-
Chromosome Location
Chr.1: 30236597 - 30239768 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015774.1 NP_001015774.1 3 - 4 396 78
GenBank mRNA BC089714.1 - 3 - 4 396 78

View Full Product Details