Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gtf2h4
Entrez Gene ID
548562
Gene Name
general transcription factor IIH subunit 4
Gene Aliases
gtf2h4.2, tfb2, tfiih
Chromosome Location
Chr.Un|NW_016686678: 1309 - 4464 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015845.1 NP_001015845.1 13 - 14 1371 63
GenBank mRNA BC090134.1 - 13 - 14 1371 63
BC122899.1 - 13 - 14 1328 63
CR848552.2 - 13 - 14 1378 63

Gene Symbol
LOC101731871
Entrez Gene ID
101731871
Gene Name
general transcription factor IIH subunit 4-like
Gene Aliases
-
Chromosome Location
Chr.2: 140338692 - 140356383 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004911912.3 XP_004911969.1 13 - 14 1422 63
XM_004911913.3 XP_004911970.1 13 - 14 1415 63
XM_004911914.3 XP_004911971.1 13 - 14 1388 63
XM_012957841.2 XP_012813295.1 13 - 14 1381 63

View Full Product Details