Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mnd1
Entrez Gene ID
548566
Gene Name
meiotic nuclear divisions 1
Gene Aliases
-
Chromosome Location
Chr.1: 45873072 - 45897729 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015849.1 NP_001015849.1 6 - 7 540 77
XM_012961773.1 XP_012817227.1 6 - 7 724 77
GenBank mRNA BC090139.1 - 6 - 7 540 77

View Full Product Details