Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
steap4
Entrez Gene ID
548569
Gene Name
STEAP4 metalloreductase
Gene Aliases
stamp2, tiarp, tnfaip9
Chromosome Location
Chr.6: 22504781 - 22518629 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015852.1 NP_001015852.1 2 - 3 985 98
XM_012964655.1 XP_012820109.1 3 - 4 1037 98
GenBank mRNA BC090357.1 - 2 - 3 985 98

View Full Product Details