Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ybx3
Entrez Gene ID
548572
Gene Name
Y-box binding protein 3
Gene Aliases
csda, csda1, dbpa, zonab
Chromosome Location
Chr.7: 10400880 - 10418766 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015855.1 NP_001015855.1 3 - 4 321 102
XM_012966546.2 XP_012822000.1 3 - 4 348 102
GenBank mRNA BC090361.1 - 3 - 4 321 102

View Full Product Details