The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015855.1 | NP_001015855.1 | 5 - 6 | 534 | 81 |
XM_012966546.2 | XP_012822000.1 | 5 - 6 | 561 | 81 | |
GenBank mRNA | BC090361.1 | - | 5 - 6 | 534 | 81 |