Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cmtm7
Entrez Gene ID
548578
Gene Name
CKLF-like MARVEL transmembrane domain containing 7
Gene Aliases
-
Chromosome Location
Chr.6: 74062177 - 74083783 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015861.1 NP_001015861.1 4 - 5 548 98
GenBank mRNA BC090372.1 - 4 - 5 548 98

View Full Product Details