Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ca7
Entrez Gene ID
548657
Gene Name
carbonic anhydrase VII
Gene Aliases
-
Chromosome Location
Chr.4: 69334380 - 69354859 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015903.1 NP_001015903.1 5 - 6 609 98
GenBank mRNA BC121632.1 - 4 - 5 439 98
BC161440.1 - 5 - 6 617 98
BC171009.1 - 5 - 6 565 98
CR926439.1 - 5 - 6 609 98

View Full Product Details