Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
msrb2
Entrez Gene ID
548659
Gene Name
methionine sulfoxide reductase B2
Gene Aliases
cbs-1, cbs1, cgi-131, pilb
Chromosome Location
Chr.6: 18216386 - 18227526 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015905.2 NP_001015905.1 2 - 3 323 79
GenBank mRNA BC170741.1 - 2 - 3 79
BC171252.1 - 2 - 3 79
CR926437.2 - 2 - 3 323 79

View Full Product Details