Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lsr
Entrez Gene ID
548689
Gene Name
lipolysis stimulated lipoprotein receptor
Gene Aliases
ildr3, lisch7, lsr.1
Chromosome Location
Chr.7: 114630341 - 114751841 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015935.2 NP_001015935.1 7 - 8 1034 69
XM_018095387.1 XP_017950876.1 6 - 7 1195 69
GenBank mRNA BC161040.1 - 8 - 9 1088 69
CR926369.2 - 7 - 8 1034 69

View Full Product Details