Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc48a1
Entrez Gene ID
548700
Gene Name
solute carrier family 48 (heme transporter), member 1
Gene Aliases
hHRG-1, hrg1, slc48a1-a, slc48a1-b
Chromosome Location
Chr.2: 141105672 - 141119727 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015946.1 NP_001015946.1 1 - 2 257 70
GenBank mRNA CR761981.2 - 1 - 2 257 70

View Full Product Details