The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001016004.2 | NP_001016004.1 | 2 - 3 | 441 | 74 |
GenBank mRNA | BC121338.1 | - | 2 - 3 | 426 | 74 |
CR848248.2 | - | 2 - 3 | 441 | 74 |