Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nup43
Entrez Gene ID
548898
Gene Name
nucleoporin 43kDa
Gene Aliases
-
Chromosome Location
Chr.5: 61842993 - 61871162 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016144.2 NP_001016144.1 3 - 4 351 83
XM_012962968.2 XP_012818422.1 3 - 4 613 83
GenBank mRNA BC161041.1 - 3 - 4 449 83
BC171085.1 - 3 - 4 351 83
CR762004.2 - 3 - 4 351 83

View Full Product Details