Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndufaf7
Entrez Gene ID
548899
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 7
Gene Aliases
c2orf56
Chromosome Location
Chr.5: 59033494 - 59050017 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016145.2 NP_001016145.1 8 - 9 1055 94
GenBank mRNA BC091018.1 - 8 - 9 1055 94
CR761995.2 - 8 - 9 968 94

View Full Product Details