Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eif3f
Entrez Gene ID
548944
Gene Name
eukaryotic translation initiation factor 3 subunit F
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683437: 157019 - 161088 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016190.2 NP_001016190.1 6 - 7 670 67
GenBank mRNA CR761801.2 - 6 - 7 670 67

View Full Product Details