Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vdac2
Entrez Gene ID
548947
Gene Name
voltage-dependent anion channel 2
Gene Aliases
-
Chromosome Location
Chr.7: 57595304 - 57618325 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016193.2 NP_001016193.1 8 - 9 783 94
XM_018095389.1 XP_017950878.1 8 - 9 929 94
GenBank mRNA BC170881.1 - 8 - 9 783 94
BC171266.1 - 8 - 9 783 94
CR761795.2 - 8 - 9 783 94

View Full Product Details