Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
samd8
Entrez Gene ID
548951
Gene Name
sterile alpha motif domain containing 8
Gene Aliases
-
Chromosome Location
Chr.7: 57640767 - 57673457 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016197.2 NP_001016197.1 3 - 4 717 103
XM_012966564.2 XP_012822018.1 3 - 4 1260 103
GenBank mRNA BC170733.1 - 2 - 3 686 103
BC170735.1 - 2 - 3 686 103
CR926348.2 - 3 - 4 717 103

View Full Product Details