Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fos
Entrez Gene ID
548954
Gene Name
FBJ murine osteosarcoma viral oncogene homolog
Gene Aliases
c-fos, xfos
Chromosome Location
Chr.8: 61168749 - 61171719 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016200.2 NP_001016200.1 1 - 2 223 79
GenBank mRNA BC123032.1 - 1 - 2 182 79
CR926345.2 - 1 - 2 223 79

View Full Product Details