Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
b9d1
Entrez Gene ID
549063
Gene Name
B9 protein domain 1
Gene Aliases
eppb9
Chromosome Location
Chr.9: 12484463 - 12513382 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016309.2 NP_001016309.1 6 - 7 589 75
XM_018097103.1 XP_017952592.1 6 - 7 523 75
GenBank mRNA BC091609.1 - 6 - 7 589 75
CR761492.2 - 6 - 7 479 75

View Full Product Details