Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rps18
Entrez Gene ID
549112
Gene Name
ribosomal protein S18
Gene Aliases
d6s218e, hke3, ke-3, ke3
Chromosome Location
Chr.8: 50971910 - 50977359 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016358.3 NP_001016358.1 3 - 4 229 67
GenBank mRNA BC135792.1 - 3 - 4 197 67
BC155978.1 - 3 - 4 196 67
BC159392.1 - 3 - 4 199 67
CN119805.1 - 3 - 4 230 67
CR761304.2 - 2 - 3 188 67

View Full Product Details