Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trappc13
Entrez Gene ID
549181
Gene Name
trafficking protein particle complex 13
Gene Aliases
c5orf44
Chromosome Location
Chr.1: 174973823 - 174997565 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016427.3 NP_001016427.2 7 - 8 620 89
NM_001244904.1 NP_001231833.1 7 - 8 620 89
GenBank mRNA BC118702.1 - 7 - 8 575 89
CR761061.2 - 7 - 8 565 89

View Full Product Details