Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
c1orf35
Entrez Gene ID
549363
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
mmtag2
Chromosome Location
Chr.6: 2077674 - 2087420 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016609.1 NP_001016609.1 4 - 5 377 88
XM_012964710.2 XP_012820164.1 5 - 6 874 88
XM_012964711.2 XP_012820165.1 5 - 6 648 88
XM_012964712.2 XP_012820166.1 5 - 6 874 88
GenBank mRNA CR760531.1 - 4 - 5 377 88

View Full Product Details