Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
elovl1
Entrez Gene ID
549398
Gene Name
ELOVL fatty acid elongase 1
Gene Aliases
-
Chromosome Location
Chr.4: 75063528 - 75082877 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016644.2 NP_001016644.1 2 - 3 107 92
GenBank mRNA CR760398.2 - 2 - 3 107 92

View Full Product Details