Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crtap
Entrez Gene ID
549401
Gene Name
cartilage associated protein
Gene Aliases
-
Chromosome Location
Chr.10: 9082448 - 9090498 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016647.2 NP_001016647.1 4 - 5 1307 89
GenBank mRNA BC135296.1 - 4 - 5 1361 89
CR760380.2 - 4 - 5 1307 89

View Full Product Details