Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snrnp25
Entrez Gene ID
549456
Gene Name
small nuclear ribonucleoprotein, U11/U12 25kDa subunit
Gene Aliases
-
Chromosome Location
Chr.9: 36942966 - 36969435 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016702.2 NP_001016702.1 5 - 6 472 81
XM_012970435.2 XP_012825889.1 5 - 6 533 81
XM_012970436.1 XP_012825890.1 5 - 6 469 81
XM_018097109.1 XP_017952598.1 5 - 6 561 81
GenBank mRNA BC091616.1 - 4 - 5 449 81
CR760143.2 - 5 - 6 472 81

View Full Product Details