Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rtn3
Entrez Gene ID
549516
Gene Name
reticulon 3
Gene Aliases
asyip, hap, nspl2, nsplii, rtn3-a, rtn3-a1, rtn3-b, xrtn3
Chromosome Location
Chr.4: 41063510 - 41087962 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016762.2 NP_001016762.1 7 - 8 628 93
XM_012953536.1 XP_012808990.1 6 - 7 1618 93
GenBank mRNA CR855761.2 - 7 - 8 628 93

View Full Product Details