Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
aqp3
Entrez Gene ID
549599
Gene Name
aquaporin 3 (Gill blood group)
Gene Aliases
AQP-x3, Aquaporin-3, aqp-3, aqp-3b, aqp3-a, aqp3-b, aqp3b, gil
Chromosome Location
Chr.1: 194788330 - 194823674 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016845.2 NP_001016845.1 3 - 4 398 92
XM_012962938.1 XP_012818392.1 4 - 5 444 92
GenBank mRNA BC158298.1 - 3 - 4 409 92
CR855446.2 - 3 - 4 398 92

View Full Product Details