Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npm2
Entrez Gene ID
549692
Gene Name
nucleophosmin/nucleoplasmin 2
Gene Aliases
-
Chromosome Location
Chr.3: 119175763 - 119180859 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016938.2 NP_001016938.1 6 - 7 612 90
GenBank mRNA CR762010.2 - 6 - 7 612 90

View Full Product Details