Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npm2
Entrez Gene ID
549692
Gene Name
nucleophosmin/nucleoplasmin 2
Gene Aliases
-
Chromosome Location
Chr.3: 119175763 - 119180859 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016938.2 NP_001016938.1 9 - 10 722 78
XM_012959066.2 XP_012814520.1 9 - 10 767 78
GenBank mRNA BC157168.1 - 9 - 10 732 78
CR762010.2 - 9 - 10 722 78

View Full Product Details