Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gata4
Entrez Gene ID
549703
Gene Name
GATA binding protein 4
Gene Aliases
gata-4, gata4-a, gata4-b, xGATA-4, xgata4
Chromosome Location
Chr.Un|NW_016683369: 772875 - 801633 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016949.2 NP_001016949.1 3 - 4 1050 82
XM_012964041.2 XP_012819495.1 4 - 5 1170 82
XM_012964042.2 XP_012819496.1 4 - 5 1273 82
GenBank mRNA BC167280.1 - 3 - 4 1043 82
CR761720.2 - 3 - 4 1050 82

View Full Product Details