Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hdgf
Entrez Gene ID
549761
Gene Name
heparin binding growth factor
Gene Aliases
hmg1l2
Chromosome Location
Chr.8: 109996941 - 110016081 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017007.2 NP_001017007.1 3 - 4 527 89
GenBank mRNA BC090580.1 - 3 - 4 527 89
CR762281.2 - 3 - 4 330 89

View Full Product Details