Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndufa4
Entrez Gene ID
549792
Gene Name
NDUFA4, mitochondrial complex associated
Gene Aliases
ci-9k, ci-mlrq, mlrq, ndufa4-a, ndufa4-b
Chromosome Location
Chr.7: 114621061 - 114623815 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017038.2 NP_001017038.1 - 168 95
GenBank mRNA BC135593.1 - - 171 95
CR761940.2 - - 168 95

View Full Product Details