Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myf6
Entrez Gene ID
549914
Gene Name
myogenic factor 6
Gene Aliases
Xmrf4, herculin, mrf4, myf6-a, myf6-b
Chromosome Location
Chr.3: 42995027 - 42999108 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017160.1 NP_001017160.1 2 - 3 597 85
XM_012959093.2 XP_012814547.1 2 - 3 85
GenBank mRNA BC153696.1 - 2 - 3 606 85
CR760833.1 - 2 - 3 597 85

View Full Product Details