Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prxl2b
Entrez Gene ID
549974
Gene Name
peroxiredoxin like 2B
Gene Aliases
c1orf93, fam213b
Chromosome Location
Chr.7: 94588230 - 94594217 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017220.2 NP_001017220.1 3 - 4 336 69
XM_012966658.2 XP_012822112.1 3 - 4 360 69
GenBank mRNA CR760360.2 - 3 - 4 336 69

View Full Product Details