Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdgfrl
Entrez Gene ID
594974
Gene Name
platelet derived growth factor receptor like
Gene Aliases
pdgrl, prlts
Chromosome Location
Chr.1: 33242605 - 33275148 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030415.2 NP_001025586.1 2 - 3 381 78
GenBank mRNA BC091030.1 - 2 - 3 382 78

View Full Product Details