Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gmfg
Entrez Gene ID
595039
Gene Name
glia maturation factor, gamma
Gene Aliases
gmf-gamma
Chromosome Location
Chr.8: 60212781 - 60215155 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030480.1 NP_001025651.1 1 - 2 474 95
GenBank mRNA BC091700.1 - 1 - 2 474 95

View Full Product Details