Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
bin3
Entrez Gene ID
595053
Gene Name
bridging integrator 3
Gene Aliases
-
Chromosome Location
Chr.3: 119633051 - 119665618 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030490.1 NP_001025661.1 6 - 7 367 68
GenBank mRNA BC092368.1 - 6 - 7 367 68

View Full Product Details