Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mettl5
Entrez Gene ID
595055
Gene Name
methyltransferase like 5
Gene Aliases
-
Chromosome Location
Chr.9: 63279151 - 63297873 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030492.1 NP_001025663.1 5 - 6 555 92
GenBank mRNA BC092545.1 - 5 - 6 555 92

View Full Product Details