Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a48
Entrez Gene ID
595080
Gene Name
solute carrier family 25 member 48
Gene Aliases
hdmcp
Chromosome Location
Chr.3: 16107090 - 16122245 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030517.1 NP_001025688.1 7 - 8 974 84
XM_012959135.1 XP_012814589.1 7 - 8 1041 84
GenBank mRNA BC094531.1 - 7 - 8 974 84

View Full Product Details