Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pm20d2
Entrez Gene ID
613074
Gene Name
peptidase M20 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 86159043 - 86175312 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032311.1 NP_001027482.1 2 - 3 657 71
GenBank mRNA BC096002.1 - 2 - 3 657 71

View Full Product Details