Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slco1b3
Entrez Gene ID
613085
Gene Name
solute carrier organic anion transporter family member 1B3
Gene Aliases
lst-3tm13, lst3, oatp1b3, oatp8, slc21a8
Chromosome Location
Chr.Un|NW_016684156: 4144 - 11735 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032322.1 NP_001027493.1 10 - 11 1922 62
GenBank mRNA BC096506.1 - 10 - 11 1922 62

View Full Product Details